(E)-4-(4-Methoxyphenyl)-4-oxobut-2-enoic acid 95%

Supplier: Ambeed
Warning

A1705253-5G A1705253-1G
77625-744EA 193.12 USD
77625-744 77876-743
(E)-4-(4-Methoxyphenyl)-4-oxobut-2-enoic acid 95%
(E)-4-(4-Methoxyphenyl)-4-oxobut-2-enoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₀O₄
Storage Temperature: Ambient
CAS Number: 20972-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR