Ifebemtinib 98%

Supplier: Ambeed
Warning

A1678539-1MG A1678539-10MG A1678539-5MG
77625-682EA 114.33 USD
77625-682 77876-664 77876-665
Ifebemtinib 98%
Ifebemtinib

Precaución: for Research Use Only

Formula: C₂₈H₂₈F₄N₆O₄
Storage Temperature: Refrigerator
MDL Number: MFCD34676862
CAS Number: 1227948-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR