GLUT inhibitor-1 98%

Supplier: Ambeed
Warning

A1602339-1MG A1602339-10MG A1602339-5MG
77625-546EA 140.82 USD
77625-546 77876-362 77876-363
GLUT inhibitor-1 98%
GLUT inhibitor-1

Precaución: for Research Use Only

Formula: C₃₂H₃₅N₇O₂
Storage Temperature: Refrigerator
MDL Number: MFCD34602693
CAS Number: 2421141-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR