Valnivudine hydrochloride 98%

Supplier: Ambeed
Warning

A1374299-1MG A1374299-5MG A1374299-10MG A1374299-100MG A1374299-25MG A1374299-50MG
77621-936EA 37.08 USD
77621-936 77621-938 77621-934 77867-698 77867-699 77867-700
Valnivudine hydrochloride 98%
Valnivudine hydrochloride

Precaución: for Research Use Only

Formula: C₂₇H₃₆ClN₃O₆
Storage Temperature: Refrigerator
MDL Number: MFCD30146050
CAS Number: 956483-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR