(-)-Alkannin 98%

Supplier: Ambeed
Warning

Synonyms: (S)-5,8-Dihydroxy-2-(1-hydroxy-4-methylpent-3-en-1-yl)naphthalene-1,4-dione, NSC 94524

A1371256-10MG A1371256-5MG
77621-890EA 40.17 USD
77621-890 77621-892
(-)-Alkannin 98%
(-)-Alkannin

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₅
MW: 288.30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00189414
CAS Number: 517-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR