Ligustrosidic acid 94%

Supplier: Ambeed
Warning

Synonyms: (E)-2-(4-(2-(4-Hydroxyphenethoxy)-2-oxoethyl)-5-(methoxycarbonyl)-2-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2H-pyran-3(4H)-ylidene)acetic acid

A1339772-10MG A1339772-5MG
77621-612EA 339.9 USD
77621-612 77621-614
Ligustrosidic acid 94%
Ligustrosidic acid

Precaución: for Research Use Only

Formula: C₂₅H₃₀O₁₄
MW: 554.50 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD34469514
CAS Number: 96382-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR