Odevixibat 98%

Supplier: Ambeed
Warning

Synonyms: A 4250, AZD 8294, A-4250, A4250, AZD-8294, (2S)-2-[[(2R)-2-[[2-[[3,3-Dibutyl-2,3,4,5-tetrahydro-7-(methylthio)-1,1-dioxido-5-phenyl-1,2,5-benzothiadiazepin-8-yl]oxy]acetyl]amino]-2-(4-hydroxyphenyl)acetyl]amino]butanoic acid, (S)-2-((R)-2-(2-((3,3-Dibutyl-7-(methylthio)-1,1-dioxido-5-phenyl-2,3,4,5-tetrahydrobenzo[f][1,2,5]thiadiazepin-8-yl)oxy)acetamido)-2-(4-hydroxyphenyl)acetamido)butanoic acid, (2S)-2-[(2R)-2-(2-{[3,3-Dibutyl-7-(methylsulfanyl)-1,1-dioxo-5-phenyl-2,3,4,5-tetrahydro-1H-1λ6,2,5-benzothiadiazepin-8-yl]oxy}acetamido)-2-(4-hydroxyphenyl)acetamido]butanoic acid, AZD8294

A1328058-10MG A1328058-5MG A1328058-100MG A1328058-25MG A1328058-50MG
77621-546EA 167.89 USD
77621-546 77621-548 77867-314 77867-315 77867-316
Odevixibat 98%
Odevixibat

Precaución: for Research Use Only

Formula: C₃₇H₄₈N₄O₈S₂
MW: 740.93 g/mol
Storage Temperature: Freezer
MDL Number: MFCD32710220
CAS Number: 501692-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR