4-Iodobutanoyl chloride 98%

Supplier: Ambeed
Danger

A1274388-5G A1274388-100MG A1274388-1G A1274388-250MG
77621-428EA 2137.25 USD
77621-428 77867-165 77867-166 77867-167
4-Iodobutanoyl chloride 98%
4-Iodobutanoyl chloride

Precaución: for Research Use Only

Formula: C₄H₆ClIO
MW: 232.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22053960
CAS Number: 927-59-3
UN: 2922
ADR: 8,6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR