Ro 41-0960 98%

Supplier: Ambeed
Warning

Synonyms: 2′-Fluoro-3,4-dihydroxy-5-nitrobenzophenone, (3,4-Dihydroxy-5-nitrophenyl)(2-fluorophenyl)methanone

A1249458-10MG A1249458-5MG
77621-230EA 157.59 USD
77621-230 77621-232
Ro 41-0960 98%
Ro 41-0960

Precaución: for Research Use Only

Formula: C₁₃H₈FNO₅
MW: 277.20 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00078597
CAS Number: 125628-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR