BGT226 95%

Supplier: Ambeed
Warning

Synonyms: NVP-BGT226, NVP-BEZ226, 8-(6-Methoxypyridin-3-yl)-3-methyl-1-(4-(piperazin-1-yl)-3-(trifluoromethyl)phenyl)-1,3-dihydro-2H-imidazo[4,5-c]quinolin-2-one

A1177233-10MG A1177233-5MG A1177233-100MG A1177233-25MG A1177233-50MG
77620-582EA 111.24 USD
77620-582 77620-584 77866-353 77866-354 77866-355
BGT226 95%
BGT226

Precaución: for Research Use Only

Formula: C₂₈H₂₅F₃N₆O₂
MW: 534.53 g/mol
MDL Number: MFCD22376639
CAS Number: 915020-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR