3,3,3-Trifluoro-2-methoxypropan-1-amine hydrochloride 95%

Supplier: Ambeed

A1130508-100MG A1130508-1G A1130508-250MG
77619-892EA 123.6 USD
77619-892 77619-894 77783-273
3,3,3-Trifluoro-2-methoxypropan-1-amine hydrochloride 95%
3,3,3-Trifluoro-2-methoxypropan-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₉ClF₃NO
MDL Number: MFCD18838805
CAS Number: 1306603-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR