Lithium tricyanomethanide - Acetonitrile Complex ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Danger
|
L0308-1G
L0308-5G
TCL0308-1GEA
157
USD
TCL0308-1G
TCL0308-5G
Lithium tricyanomethanide - Acetonitrile Complex ≥98.0% (by titrimetric analysis)
Lithium tricyanomethanide - Acetonitrile Complex
Formula:
C₄LiN₃·CH₃CN MW: 138.06 g/mol |
UN:
3439 ADR: 6.1,II |
Specification Test Results
Appearance | White to Almost white powder to crystaline |
Purity (Ion exchange titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...