trans,trans-4-Butyl-4'-vinylbicyclohexyl ≥98.0% (by GC)

Supplier: TCI America

B5123-1G B5123-5G
TCB5123-1GEA 32 USD
TCB5123-1G TCB5123-5G
trans,trans-4-Butyl-4'-vinylbicyclohexyl ≥98.0% (by GC)
trans,trans-4-Butyl-4'-vinylbicyclohexyl
Formula: C₁₈H₃₂
MDL Number: MFCD25541699
CAS Number: 153429-47-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.000 %
Melting point 53.0 to 57.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR