trans,trans-4-Pentyl-4'-vinylbicyclohexyl ≥98.0% (by GC)

Supplier: TCI America

Synonyms: trans,trans-4-Amyl-4'-vinylbicyclohexyl

P2318-1G P2318-5G
TCP2318-1GEA 34.5 USD
TCP2318-1G TCP2318-5G
trans,trans-4-Pentyl-4'-vinylbicyclohexyl ≥98.0% (by GC)
trans,trans-4-Pentyl-4'-vinylbicyclohexyl
Formula: C₁₉H₃₄
MDL Number: MFCD23135326
CAS Number: 129738-34-7

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Melting point 47.0 to 51.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR