3-Amino-1-benzylpiperidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
A2917-1G
A2917-5G
TCA2917-1GEA
84
USD
TCA2917-1G
TCA2917-5G
3-Amino-1-benzylpiperidine ≥98.0% (by GC, titration analysis)
3-Amino-1-benzylpiperidine
Formula:
C₁₂H₁₈N₂ Boiling Pt: 82°C Storage Temperature: Refrigerator |
MDL Number:
MFCD04038444 CAS Number: 60407-35-4 |
Specification Test Results
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...