Ethyl 4-Fluorocinnamate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Fluorocinnamic acid ethyl ester, Ethyl (2E)-3-(4-fluorophenyl)acrylate

E1099-1G E1099-5G
TCE1099-1GEA 243 USD
TCE1099-1G TCE1099-5G
Ethyl 4-Fluorocinnamate ≥98.0% (by GC)
Ethyl 4-Fluorocinnamate
Formula: C₁₁H₁₁FO₂
MW: 194.21 g/mol
Boiling Pt: 131°C
Storage Temperature: Refrigerator
MDL Number: MFCD02252626
CAS Number: 352-03-4

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR