Hexafluoroisopropyl methyl ether ≥98.0% (by GC)
Supplier: TCI America
|
Danger
|
H1524-25G
H1524-5G
TCH1524-25GEA
310.7
USD
TCH1524-25G
TCH1524-5G
Hexafluoroisopropyl methyl ether ≥98.0% (by GC)
Hexafluoroisopropyl methyl ether
Formula:
C₄H₄F₆O Boiling Pt: 50°C |
MDL Number:
MFCD00221772 CAS Number: 22052-84-2 UN: 3271 ADR: 3,II |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...