6,7-Dimethoxy-3,4-dihydroisoquinoline hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

D4629-1G D4629-5G
TCD4629-1GEA 29 USD
TCD4629-1G TCD4629-5G
6,7-Dimethoxy-3,4-dihydroisoquinoline hydrochloride ≥98.0% (by HPLC, titration analysis)
6,7-Dimethoxy-3,4-dihydroisoquinoline hydrochloride
Formula: C₁₁H₁₄ClNO₂
MW: 227.69 g/mol
MDL Number: MFCD00143492
CAS Number: 20232-39-7

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 192.0 to 196.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR