1-Methyl-3-propylimidazolium bis(trifluoromethylsulphonyl)imide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
1-Methyl-3-propylimidazolium Bis(trifluoromethanesulphonyl)imide
M3059-25G
M3059-5G
76220-344EA
198
USD
76220-344
76220-346
1-Methyl-3-propylimidazolium bis(trifluoromethylsulphonyl)imide ≥98.0% (by HPLC, titration analysis)
1-Methyl-3-propylimidazolium bis(trifluoromethylsulphonyl)imide
Formula:
C₉H₁₃F₆N₃O₄S₂ MW: 405.34 g/mol |
MDL Number:
MFCD15071469 CAS Number: 216299-72-8 |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (HPLC) | min. 98.0 area% |
Purity (Ion exchange titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...