(2-Bromoethyl)phosphonic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

B5013-1G B5013-200MG
TCB5013-1GEA 490.6 USD
TCB5013-1G TCB5013-200MG
(2-Bromoethyl)phosphonic acid ≥98.0% (by titrimetric analysis)
(2-Bromoethyl)phosphonic acid
Formula: C₂H₆BrO₃P
MDL Number: MFCD00013957
CAS Number: 999-82-6
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 86.0 to 90.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR