5-Azoniaspiro[4.4]nonane chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
A3092-1G
A3092-5G
TCA3092-1GEA
70
USD
TCA3092-1G
TCA3092-5G
5-Azoniaspiro[4.4]nonane chloride ≥98.0% (by titrimetric analysis)
5-Azoniaspiro[4.4]nonane chloride
Formula:
C₈H₁₆ClN Melting Pt: 295°C |
CAS Number:
98997-63-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...