N,N'-Ditridecyl-3,4,9,10-perylenetetracarboxylic diimide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: PTCDI-C13

D3947-1G D3947-200MG
TCD3947-1GEA 198 USD
TCD3947-1G TCD3947-200MG
N,N'-Ditridecyl-3,4,9,10-perylenetetracarboxylic diimide ≥98.0% (by HPLC, total nitrogen)
N,N'-Ditridecyl-3,4,9,10-perylenetetracarboxylic diimide
Formula: C₅₀H₆₂N₂O₄
MW: 755.06 g/mol
MDL Number: MFCD00134587
CAS Number: 95689-92-2

Order Now

Specification Test Results

Appearance Orange to Brown to Dark red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR