1-Bromo-4-(4-bromophenyl)naphthalene ≥97.0% (by GC)

Supplier: TCI America

B5056-1G B5056-5G
76218-798EA 161 USD
76218-798 76218-800
1-Bromo-4-(4-bromophenyl)naphthalene ≥97.0% (by GC)
1-Bromo-4-(4-bromophenyl)naphthalene
Formula: C₁₆H₁₀Br₂
CAS Number: 952604-26-1

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 146.0 to 150.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR