N,N-Di(4-biphenylyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
4-[Di(4-biphenylyl)amino]phenylboronic acid Pinacol ester, 2-[4-[Di(4-biphenylyl)amino]phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, N-([1,1'-Biphenyl]-4-yl)-N-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-[1,1'-biphenyl]-4-amine
D4886-1G
D4886-5G
TCD4886-1GEA
170
USD
TCD4886-1G
TCD4886-5G
N,N-Di(4-biphenylyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline ≥98.0% (by HPLC, titration analysis)
N,N-Di(4-biphenylyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Formula:
C₃₆H₃₄BNO₂ MW: 523.48 g/mol |
MDL Number:
MFCD29089353 CAS Number: 952431-30-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...