9-(6-Bromohexyl)-9H-carbazole ≥97.0% (by GC)
Supplier: TCI America
|
Warning
|
B5548-1G
B5548-200MG
TCB5548-1GEA
269
USD
TCB5548-1G
TCB5548-200MG
9-(6-Bromohexyl)-9H-carbazole ≥97.0% (by GC)
9-(6-Bromohexyl)-9H-carbazole
Formula:
C₁₈H₂₀BrN |
MDL Number:
MFCD30562129 CAS Number: 94847-10-6 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 58.0 to 62.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...