3,6-Dibromo-9-[4-(2-ethylhexyloxy)phenyl]-9H-carbazole ≥95.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

D5342-1G D5342-200MG
TCD5342-1GEA 245 USD
TCD5342-1G TCD5342-200MG
3,6-Dibromo-9-[4-(2-ethylhexyloxy)phenyl]-9H-carbazole ≥95.0% (by HPLC, total nitrogen)
3,6-Dibromo-9-[4-(2-ethylhexyloxy)phenyl]-9H-carbazole
Formula: C₂₆H₂₇Br₂NO
MDL Number: MFCD30721573
CAS Number: 946491-48-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR