Bis(4-bromophenyl)phenylphosphine oxide ≥98.0% (by GC)

Supplier: TCI America
Warning

B5532-1G B5532-200MG
TCB5532-1GEA 182 USD
TCB5532-1G TCB5532-200MG
Bis(4-bromophenyl)phenylphosphine oxide ≥98.0% (by GC)
Bis(4-bromophenyl)phenylphosphine oxide
Formula: C₁₈H₁₃Br₂OP
MW: 436.08 g/mol
Boiling Pt: 280°C
CAS Number: 93869-52-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 105.0 to 109.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR