9-(4-Bromophenyl)-9-phenylfluorene ≥98.0% (by GC)

Supplier: TCI America

B5221-1G B5221-5G
TCB5221-1GEA 60 USD
TCB5221-1G TCB5221-5G
9-(4-Bromophenyl)-9-phenylfluorene ≥98.0% (by GC)
9-(4-Bromophenyl)-9-phenylfluorene
Formula: C₂₅H₁₇Br
MDL Number: MFCD28975087
CAS Number: 937082-81-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 151.0 to 155.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR