3-Bromo-9-(2-naphthyl)carbazole ≥97.0% (by HPLC, total nitrogen)
Supplier: TCI America
|
Warning
|
B5028-1G
B5028-5G
TCB5028-1GEA
61
USD
TCB5028-1G
TCB5028-5G
3-Bromo-9-(2-naphthyl)carbazole ≥97.0% (by HPLC, total nitrogen)
3-Bromo-9-(2-naphthyl)carbazole
Formula:
C₂₂H₁₄BrN |
MDL Number:
MFCD26127422 CAS Number: 934545-80-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...