1-Methyl-1-n-octylpyrrolidinium bis(trifluoromethanesulfonyl)imide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

M3117-5G M3117-25G
TCM3117-5GEA 160 USD
TCM3117-5G TCM3117-25G
1-Methyl-1-n-octylpyrrolidinium bis(trifluoromethanesulfonyl)imide ≥98.0% (by titrimetric analysis)
1-Methyl-1-n-octylpyrrolidinium bis(trifluoromethanesulfonyl)imide
Formula: C₁₅H₂₈F₆N₂O₄S₂
Melting Pt: -12°C
MDL Number: MFCD09038851
CAS Number: 927021-43-0

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR