N,N-Dimethylpropan-1-amine ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: N,N-Dimethyl-1-propanamine, N,N-Dimethyl Propylamine

D5821-25ML D5821-5ML
TCD5821-25MLEA 249 USD
TCD5821-25ML TCD5821-5ML
N,N-Dimethylpropan-1-amine ≥98.0% (by GC)
N,N-Dimethylpropan-1-amine
Formula: C₅H₁₃N
MW: 87.17 g/mol
Boiling Pt: 66°C
MDL Number: MFCD00015203
CAS Number: 926-63-6
EINECS: 213-139-9
UN: 2266
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Specific gravity (20/20) 0.7000 to 0.7100

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR