N-[2-(Dimethylamino)ethyl]acrylamide ≥98.0% (by GC, titration analysis) stabilized

Supplier: TCI America
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

D5794-25G D5794-5G
TCD5794-25GEA 615 USD
TCD5794-25G TCD5794-5G
N-[2-(Dimethylamino)ethyl]acrylamide ≥98.0% (by GC, titration analysis) stabilized
N-[2-(Dimethylamino)ethyl]acrylamide
Formula: C₇H₁₄N₂O
Boiling Pt: 125°C
Storage Temperature: Refrigerator
MDL Number: MFCD00053694
CAS Number: 925-76-8
EINECS: 213-124-7

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Specific gravity (20/20) 0.9600 to 0.9700


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR