2-Bromo-8-iododibenzofuran ≥98.0% (by GC)

Supplier: TCI America

B4867-1G B4867-200MG
TCB4867-1GEA 276.2 USD
TCB4867-1G TCB4867-200MG
2-Bromo-8-iododibenzofuran ≥98.0% (by GC)
2-Bromo-8-iododibenzofuran
Formula: C₁₂H₆BrIO
MW: 372.99 g/mol
MDL Number: MFCD28975101
CAS Number: 916435-41-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 171.0 to 175.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR