2,8-Bis(9H-carbazol-9-yl)dibenzothiophene ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
DCzDBT, 2,8-Di(9-carbazolyl)dibenzothiophene
B4942-1G
B4942-200MG
TCB4942-1GEA
241.7
USD
TCB4942-1G
TCB4942-200MG
2,8-Bis(9H-carbazol-9-yl)dibenzothiophene ≥98.0% (by HPLC, total nitrogen)
2,8-Bis(9H-carbazol-9-yl)dibenzothiophene
Formula:
C₃₆H₂₂N₂S MW: 514.65 g/mol |
MDL Number:
MFCD28291905 CAS Number: 913738-04-2 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...