4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzofuran ≥99.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-(Dibenzo[b,d]furan-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, Dibenzofuran-4-boronic acid Pinacol ester, 2-(Dibenzofuran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

T3602-1G T3602-5G
TCT3602-1GEA 103 USD
TCT3602-1G TCT3602-5G
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzofuran ≥99.0% (by GC, titration analysis)
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzofuran
Formula: C₁₈H₁₉BO₃
MDL Number: MFCD18910632
CAS Number: 912824-85-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %
Melting point 107.0 to 111.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR