1-Methyl-3-(4-sulfobutyl)imidazolium bis(trifluoromethanesulfonyl)imide ≥95.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
M3119-1G
M3119-5G
TCM3119-1GEA
68
USD
TCM3119-1G
TCM3119-5G
1-Methyl-3-(4-sulfobutyl)imidazolium bis(trifluoromethanesulfonyl)imide ≥95.0% (by titrimetric analysis)
1-Methyl-3-(4-sulfobutyl)imidazolium bis(trifluoromethanesulfonyl)imide
Formula:
C₁₀H₁₅F₆N₃O₇S₃ |
CAS Number:
909390-59-6 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...