(4-Nitrophenyl)(phenyl)iodonium trifluoromethanesulfonate ≥98.0% (by HPLC)
Supplier: TCI America
|
Warning
|
Synonyms:
(4-Nitrophenyl)(phenyl)iodonium triflate
N1066-1G
N1066-5G
76220-376EA
219
USD
76220-376
76220-378
(4-Nitrophenyl)(phenyl)iodonium trifluoromethanesulfonate ≥98.0% (by HPLC)
(4-Nitrophenyl)(phenyl)iodonium trifluoromethanesulfonate
Formula:
C₁₃H₉F₃INO₅S MW: 475.18 g/mol |
MDL Number:
MFCD21608480 CAS Number: 905718-45-8 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Ion exchange titration) | min. 97.0 % |
Melting point | 198.0 to 202.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...