9-(2-Bromophenyl)-9H-carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

B4109-1G B4109-5G
TCB4109-1GEA 29 USD
TCB4109-1G TCB4109-5G
9-(2-Bromophenyl)-9H-carbazole ≥98.0% (by GC)
9-(2-Bromophenyl)-9H-carbazole
Formula: C₁₈H₁₂BrN
MW: 322.21 g/mol
MDL Number: MFCD23135883
CAS Number: 902518-11-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 99.0 to 103.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR