Xylan (from corn core)

Supplier: TCI America

X0078-100G X0078-25G
TCX0078-100GEA 299.6 USD
TCX0078-100G TCX0078-25G
Xylan (from corn core)
Xylan
MDL Number: MFCD00082148
CAS Number: 9014-63-5
EINECS: 232-760-6

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Xylose min. 75.0 % (after hydrolysis)
Drying loss max. 15.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR