1,4-Dibromo-2-iodobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

D4906-1G D4906-5G
76219-750EA 23 USD
76219-750 76219-752
1,4-Dibromo-2-iodobenzene ≥98.0% (by GC)
1,4-Dibromo-2-iodobenzene
Formula: C₆H₃Br₂I
Boiling Pt: 180°C
MDL Number: MFCD07778993
CAS Number: 89284-52-6

Order Now

Specification Test Results

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 39.0 to 43.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR