3,3'-Methylenebis[1-(2,6-diisopropylphenyl)-3-imidazolium bromide] ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

M2791-1G M2791-5G
TCM2791-1GEA 228 USD
TCM2791-1G TCM2791-5G
3,3'-Methylenebis[1-(2,6-diisopropylphenyl)-3-imidazolium bromide] ≥98.0% (by HPLC, titration analysis)
3,3'-Methylenebis[1-(2,6-diisopropylphenyl)-3-imidazolium bromide]
Formula: C₃₁H₄₂Br₂N₄
MW: 630.51 g/mol
Melting Pt: 277°C
CAS Number: 889617-36-1

Order Now

Specification Test Results

Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR