Liranaftate ≥98.0% (by HPLC)

Supplier: TCI America
Warning

L0301-250MG L0301-50MG
TCL0301-250MGEA 686 USD
TCL0301-250MG TCL0301-50MG
Liranaftate ≥98.0% (by HPLC)
Liranaftate
Formula: C₁₈H₂₀N₂O₂S
MDL Number: MFCD00865576
CAS Number: 88678-31-3
Merck Index: 14,05515

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 8.00 to 9.00 %
Melting point 97.0 to 101.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR