2,7-Di(1-pyrenyl)-9,9'-spirobi[9H-fluorene] ≥98.0% (by HPLC)

Supplier: TCI America

D4932-1G D4932-200MG
76219-762EA 395 USD
76219-762 76219-764
2,7-Di(1-pyrenyl)-9,9'-spirobi[9H-fluorene] ≥98.0% (by HPLC)
2,7-Di(1-pyrenyl)-9,9'-spirobi[9H-fluorene]
Formula: C₅₇H₃₂
MDL Number: MFCD12022456
CAS Number: 886456-80-0

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR