4-(trans-4-Butylcyclohexyl)phenol ≥98.0% (by GC)

Supplier: TCI America

B2644-100G B2644-25G
TCB2644-100GEA 256 USD
TCB2644-100G TCB2644-25G
4-(trans-4-Butylcyclohexyl)phenol ≥98.0% (by GC)
4-(trans-4-Butylcyclohexyl)phenol
Formula: C₁₆H₂₄O
MDL Number: MFCD00673752
CAS Number: 88581-00-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR