3-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Methylthiophene-2-boronic acid pinacol ester, 4,4,5,5-Tetramethyl-2-(3-methylthiophen-2-yl)-1,3,2-dioxaborolane, 3-Methyl-2-thiopheneboronic acid Pinacol ester, 2-(3-Methyl-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

M2391-1G M2391-5G
TCM2391-1GEA 92 USD
TCM2391-1G TCM2391-5G
3-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥98.0% (by GC, titration analysis)
3-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene
Formula: C₁₁H₁₇BO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD08063137
CAS Number: 885692-91-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 58.0 to 62.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR