5-Formyl-2-methoxyphenyl Acetate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Acetic acid 5-Formyl-2-methoxyphenyl ester, 3-Acetoxy-4-methoxybenzaldehyde

A2946-25G A2946-5G
TCA2946-25GEA 388.1 USD
TCA2946-25G TCA2946-5G
5-Formyl-2-methoxyphenyl Acetate ≥98.0% (by GC)
5-Formyl-2-methoxyphenyl Acetate
Formula: C₁₀H₁₀O₄
MDL Number: MFCD00266441
CAS Number: 881-57-2

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 85.0 to 89.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR