9-Phenyl-9H-carbazole-3-carboxaldehyde ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 9-Phenyl-9H-carbazole-3-carbaldehyde, 3-Formyl-9-phenyl-9H-carbazole

P2351-1G P2351-5G
TCP2351-1GEA 179 USD
TCP2351-1G TCP2351-5G
9-Phenyl-9H-carbazole-3-carboxaldehyde ≥98.0% (by GC)
9-Phenyl-9H-carbazole-3-carboxaldehyde
Formula: C₁₉H₁₃NO
MW: 271.32 g/mol
MDL Number: MFCD19441087
CAS Number: 87220-68-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 104.0 to 108.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR