2,6-Bis(diphenylamino)anthraquinone ≥96.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 2,6-Bis(diphenylamino)anthracene-9,10-dione

B5092-1G B5092-200MG
76218-810EA 281 USD
76218-810 76218-812
2,6-Bis(diphenylamino)anthraquinone ≥96.0% (by HPLC, total nitrogen)
2,6-Bis(diphenylamino)anthraquinone
Formula: C₃₈H₂₆N₂O₂
MW: 542.64 g/mol
MDL Number: MFCD20040460
CAS Number: 868850-50-4

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (with Total Nitrogen) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR