Polyethylene Glycol Glycidyl Lauryl ether
Supplier: TCI America
P2294-25G
P2294-500G
TCP2294-25GEA
48
USD
TCP2294-25G
TCP2294-500G
Polyethylene Glycol Glycidyl Lauryl ether
Polyethylene Glycol Glycidyl Lauryl ether
CAS Number:
86630-59-3 |
Specification Test Results
Appearance | White to Almost white cloudy liquid |
Epoxy equivalent | 900 to 1,100 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...