3,6-Dibromo-9-(4-dodecyloxyphenyl)-9H-carbazole ≥97.0% (by HPLC, total nitrogen)
Supplier: TCI America
|
Warning
|
D5343-1G
D5343-200MG
TCD5343-1GEA
275
USD
TCD5343-1G
TCD5343-200MG
3,6-Dibromo-9-(4-dodecyloxyphenyl)-9H-carbazole ≥97.0% (by HPLC, total nitrogen)
3,6-Dibromo-9-(4-dodecyloxyphenyl)-9H-carbazole
Formula:
C₃₀H₃₅Br₂NO MW: 585.42 g/mol |
MDL Number:
MFCD30536722 CAS Number: 865163-47-9 |
Specification Test Results
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 89.0 to 93.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...